• USA Home
  • N8518 - Naphthol AS-TR phosphate disodium salt

EMAIL THIS PAGE TO A FRIEND
N8518 Sigma-Aldrich

Naphthol AS-TR phosphate disodium salt

tablet, 4 mg substrate per tablet

  • CAS Number 4264-93-1

  • Empirical Formula (Hill Notation) C18H13ClNNa2O5P

  • Molecular Weight 435.71

  •  MDL number MFCD00069550

  •  PubChem Substance ID 24897683

Purchase

Description

Specificity

Naphthol AS-TR Phosphate is a substrate for use with alkaline phosphatase in immunoblotting and immunohistological staining procedures. When Naphthol AS-TR Phosphate is used in conjunction with Fast Red RC Tablets (Sigma Product No. F 5146) an insoluble end product is produced that is red in color and can be observed visually. Naphthol AS-TR Phosphate Tablets may also be used with other coupling salts in other applications.

Application

Substrate for the histochemical demonstration of acid and alkaline phosphatase.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Colorimetric Alkaline Phosphatase and Peroxidase Substrate Detection Systems

Nitroblue Tetrazolium (NBT) is used with the alkaline phosphatase substrate 5-Bromo- 4-Chloro-3-Indolyl Phosphate (BCIP) in western blotting and immunohistological staining procedures. These substrat...
BioFiles 2008, 3.4, 6.
Keywords: Enzyme-linked immunosorbent assay, Immunohistochemistry, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?