• USA Home
  • N8763 - NBD-Propranolol dihydrochloride

EMAIL THIS PAGE TO A FRIEND
N8763 Sigma-Aldrich

NBD-Propranolol dihydrochloride

Synonym: N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)propranolol

  • CAS Number 108321-35-3

  • Empirical Formula (Hill Notation) C21H21N5O5 · 2HCl

  • Molecular Weight 496.34

Purchase

Properties

storage temp.   −20°C

Description

Application

Fluorescent β-receptor probe

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?