• USA Home
  • N9009 - 5-Nitro-2-furaldehyde semicarbazone

EMAIL THIS PAGE TO A FRIEND
N9009 Sigma-Aldrich

5-Nitro-2-furaldehyde semicarbazone

Synonym: 5-Nitro-2-furfural semicarbazone, Furacin, NFZ, Nitrofurazone

  • CAS Number 59-87-0

  • Empirical Formula (Hill Notation) C6H6N4O4

  • Molecular Weight 198.14

  •  Beilstein/REAXYS Number 86403

  •  EC Number 200-443-1

  •  MDL number MFCD00003225

Purchase

Properties

InChI Key   IAIWVQXQOWNYOU-FPYGCLRLSA-N
mp   242-244 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LT7700000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Carcinogen Selector - Endocrine System - Mammary Gland

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Ovary Endocrine

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Ovary

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Reproductive System
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?