EMAIL THIS PAGE TO A FRIEND
O1381 Supelco

Oxazepam-d5

Synonym: 7-Chloro-1,3-dihydro-3-hydroxy-5-(phenyl-d5)-2H-1,4-benzodiazepin-2-one

Purchase

Properties

InChI Key   ADIMAYPTOBDMTL-RALIUCGRSA-N
form   neat
drug control   USDEA Schedule IV; Home Office Schedule 4.1; psychotrope; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
mass shift   M+5
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

O5129 Oxazepam-d5

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?