EMAIL THIS PAGE TO A FRIEND
O4502 Sigma

Oxypurinol

≥98% (HPLC)

Purchase

Properties

InChI Key   HXNFUBHNUDHIGC-UHFFFAOYSA-N
assay   ≥98% (HPLC)

Description

Biochem/physiol Actions

An allopurinol metabolite.

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

O6881 Oxypurinol, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?