EMAIL THIS PAGE TO A FRIEND
O4754 Sigma-Aldrich

Methyl oleate

≥99%

Synonym: Methyl cis-9-octadecenoate, Oleic acid methyl ester

  • CAS Number 112-62-9

  • Linear Formula CH3(CH2)7CH=CH(CH2)7CO2CH3

  • Molecular Weight 296.49

  •  Beilstein/REAXYS Number 1727037

  •  EC Number 203-992-5

  •  MDL number MFCD00009578

Purchase

Properties

vapor pressure   10 mmHg ( 205 °C)
assay   ≥99%
refractive index   n20/D 1.452 (lit.)
bp   218 °C/20 mmHg (lit.)
density   0.874 g/mL at 20 °C (lit.)
storage temp.   −20°C
SMILES string   CCCCCCCC\C=C/CCCCCCCC(=O)OC
InChI   1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10-
InChI key   QYDYPVFESGNLHU-KHPPLWFESA-N

Description

Packaging

Sealed ampule.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
RK0895000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?