EMAIL THIS PAGE TO A FRIEND
O4875 Sigma-Aldrich

(+)-Octopine

Synonym: N2-(1-Carboxyethyl)-L-arginine, N2-([R]-1-Carboxyethyl)-L-arginine

  • CAS Number 34522-32-2

  • Linear Formula H2NC(=NH)NH(CH2)3CH[NHCH(CH3)CO2H]CO2H

  • Molecular Weight 246.26

  •  MDL number MFCD00078080

  •  PubChem Substance ID 24898005

Purchase

Properties

mp   283-284 °C (lit.)
storage temp.   −20°C
SMILES string   C[C@@H](N[C@@H](CCCNC(N)=N)C(O)=O)C(O)=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?