• USA Home
  • O4880 - cis-6,9,12,15-Octadecatetraenoic acid

EMAIL THIS PAGE TO A FRIEND
O4880 Sigma-Aldrich

cis-6,9,12,15-Octadecatetraenoic acid

~90% (capillary GC)

Synonym: Stearidonic acid

  • CAS Number 2091-28-3

  • Empirical Formula (Hill Notation) C18H28O2

  • Molecular Weight 276.41

  •  MDL number MFCD00216036

Purchase

Properties

assay   ~90% (capillary GC)
storage temp.   −20°C
SMILES string   CC\C=C\C\C=C\C\C=C\C\C=C\CCCCC(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?