EMAIL THIS PAGE TO A FRIEND
O5000 Sigma-Aldrich

(+)-Octopinic acid

Synonym: N2-([R]-1-Carboxyethyl)-L-ornithine

  • CAS Number 20197-09-5

  • Empirical Formula (Hill Notation) C8H16N2O4

  • Molecular Weight 204.22

  •  MDL number MFCD00057635

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?