EMAIL THIS PAGE TO A FRIEND
O5255 Sigma

cis-3-Octen-1-ol

~96%

Purchase

Properties

assay   ~96%
refractive index   n20/D 1.447(lit.)
bp   73 °C/0.5 mmHg(lit.)
density   0.849 g/mL at 25 °C(lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NA 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?