• USA Home
  • O5750 - Oxytetracycline dihydrate

EMAIL THIS PAGE TO A FRIEND
O5750 Sigma-Aldrich

Oxytetracycline dihydrate

crystalline, ≥97%

Purchase

Properties

Related Categories Antibacterial, Antibiotics, Antibiotics A to Z, Antibiotics N-S, Antibiotics by Application,
InChI Key   IBZHEBHGZFICKS-IFLJXUKPSA-N
assay   ≥97%
form   crystalline
color   yellow
Mode of action   protein synthesis | interferes
antibiotic activity spectrum   Gram-negative bacteria
  Gram-positive bacteria

Description

General description

Chemical structure: tetracycline

Biochem/physiol Actions

Antibiotic produced by Streptomyces rimosus.
Mode of Action: Inhibits protein synthesis (elongation) by preventing binding of aminoacyl-tRNA to the 30S subunit.
Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.
Mode of Resistance: Active efflux, ribosome protection, tetracycline inactivation.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

O4636 Oxytetracycline dihydrate, Assay (anhydrous basis) 95.0%-102.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?