• USA Home
  • O5756 - Octyl β-D-thiogalactopyranoside

EMAIL THIS PAGE TO A FRIEND
O5756 Sigma-Aldrich

Octyl β-D-thiogalactopyranoside

  • CAS Number 42891-16-7

  • Empirical Formula (Hill Notation) C14H28O5S

  • Molecular Weight 308.43

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?