• USA Home
  • P0208 - Pro-Gln-Arg-Phe amide trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
P0208 Sigma-Aldrich

Pro-Gln-Arg-Phe amide trifluoroacetate salt

≥97% (HPLC)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Opioid Peptides, Other Opioid Peptides, Peptides and Proteins,
assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   NC(=N)NCCCC(NC(=O)C(CCC(N)=O)NC(=O)C1CCCN1)C(=O)NC(Cc2ccccc2)C(N)=O
InChI   1S/C25H39N9O5/c26-20(35)11-10-18(33-22(37)16-8-4-12-30-16)24(39)32-17(9-5-13-31-25(28)29)23(38)34-19(21(27)36)14-15-6-2-1-3-7-15/h1-3,6-7,16-19,30H,4-5,8-14H2,(H2,26,35)(H2,27,36)(H,32,39)(H,33,37)(H,34,38)(H4,28,29,31)
InChI key   VVKYIUFJEDANQO-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Morphine-modulating peptide

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?