EMAIL THIS PAGE TO A FRIEND
P0260 Sigma-Aldrich

Palmityl acetate

≥99%

Purchase

Properties

assay   ≥99%
form   solid
storage temp.   −20°C
Gene Information   human ... CNR1(1268), CNR2(1269)
rat ... Faah(29347)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?