• USA Home
  • P0654 - Ethyl β-carboline-3-carboxylate

EMAIL THIS PAGE TO A FRIEND
P0654 Sigma-Aldrich

Ethyl β-carboline-3-carboxylate

~95%

Synonym: β-CCE, Ethyl 9H-pyrido[3,4-b]indole-3-carboxylate

  • CAS Number 74214-62-3

  • Empirical Formula (Hill Notation) C14H12N2O2

  • Molecular Weight 240.26

  •  MDL number MFCD00009730

Purchase

Properties

assay   ~95%
mp   228-230 °C (lit.)
SMILES string   CCOC(=O)c1cc2c(cn1)[nH]c3ccccc23
InChI   1S/C14H12N2O2/c1-2-18-14(17)12-7-10-9-5-3-4-6-11(9)16-13(10)8-15-12/h3-8,16H,2H2,1H3
InChI key   KOVRZNUMIKACTB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RN7092000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?