EMAIL THIS PAGE TO A FRIEND
P0760 Sigma-Aldrich

Palmitic anhydride

Grade I, ~99%

Synonym: Hexadecanoic anhydride

  • CAS Number 623-65-4

  • Linear Formula [CH3(CH2)14CO]2O

  • Molecular Weight 494.83

  •  Beilstein/REAXYS Number 1807507

  •  EC Number 210-805-0

  •  MDL number MFCD00008992

Purchase

Properties

InChI Key   QWZBEFCPZJWDKC-UHFFFAOYSA-N
type   Grade I
assay   ~99%
mp   61-64 °C (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

286508 Palmitic anhydride, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?