• USA Home
  • P0815 - Prostaglandin E1 ethyl ester

EMAIL THIS PAGE TO A FRIEND
P0815 Sigma-Aldrich

Prostaglandin E1 ethyl ester

~5 mg/mL in methyl acetate, ~98% (HPLC)

Purchase

Properties

assay   ~98% (HPLC)
concentration   ~5 mg/mL in methyl acetate
storage temp.   −20°C

Description

Biochem/physiol Actions

Ethyl ester form of prostaglandin E1, a vasodilator that activates adenylyl cyclase. Ethyl ester PGE1 converts to PGE1 by enzymatic hydrolysis.

Packaging

Packaged under Argon.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1231 - class 3 - PG 2 - Methyl acetate, solution
WGK Germany 
2
Flash Point(F) 
33.8 °F
Flash Point(C) 
1 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?