• USA Home
  • P1023 - 5β-Pregnane-3α,11β,20β-triol

EMAIL THIS PAGE TO A FRIEND
P1023 Supelco

5β-Pregnane-3α,11β,20β-triol

Synonym: 3α,11β,20β-Trihydroxy-5β-pregnane

  • CAS Number 10417-85-3

  • Empirical Formula (Hill Notation) C21H36O3

  • Molecular Weight 336.51

Purchase
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?