• USA Home
  • P1125 - Phe-Leu amide hydrochloride

EMAIL THIS PAGE TO A FRIEND
P1125 Sigma-Aldrich

Phe-Leu amide hydrochloride

  • CAS Number 81638-86-0

  • Empirical Formula (Hill Notation) C15H23N3O2 · HCl

  • Molecular Weight 313.82

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?