• USA Home
  • P1293 - 2-Arachidonoyl-1-stearoyl-sn-glycerol 3-phosphate

EMAIL THIS PAGE TO A FRIEND
P1293 Sigma-Aldrich

2-Arachidonoyl-1-stearoyl-sn-glycerol 3-phosphate

10 mg/mL in chloroform, ~97% (positional purity of fatty acids), ~98% (TLC)

Synonym: L-α-Phosphatidic acid, β-arachidonoyl-γ-stearoyl sodium salt

  • CAS Number 64665-00-5

  • Empirical Formula (Hill Notation) C41H73O8P

  • Molecular Weight 724.99

Purchase

Properties

assay   ~97% (positional purity of fatty acids)
  ~98% (TLC)
concentration   10 mg/mL in chloroform
shipped in   dry ice
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
UN 1888 6.1/PG 3
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?