• USA Home
  • P1301 - Polyoxyethylene (7) stearyl ether

EMAIL THIS PAGE TO A FRIEND
P1301 Sigma-Aldrich

Polyoxyethylene (7) stearyl ether

~95%

Synonym: C18E7, Heptaethylene glycol monooctadecyl ether, Octadecylheptaglycol

  • CAS Number 66146-84-7

  • Linear Formula CH3(CH2)17(OCH2CH2)7OH

  • Molecular Weight 578.86

  •  MDL number MFCD00077929

Purchase

Properties

description   non-ionic
assay   ~95%
SMILES string   CCCCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCO
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?