• USA Home
  • P131 - Phentolamine methanesulfonate salt

EMAIL THIS PAGE TO A FRIEND
P131 Sigma-Aldrich

Phentolamine methanesulfonate salt

Synonym: 2-(N-[m-Hydroxyphenyl]-p-toluidinomethyl)imidazoline

Purchase

Properties

InChI Key   OGIYDFVHFQEFKQ-UHFFFAOYSA-N
mp   180-182 °C (lit.)

Description

Biochem/physiol Actions

α-Adrenergic receptor antagonist; peripheral vasodilator.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SL5430000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P7561 Phentolamine methanesulfonate salt, ≥98% (TLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?