• USA Home
  • P1648 - 5β-Pregnane-3β,11β,17α,21-tetrol-20-one

EMAIL THIS PAGE TO A FRIEND
P1648 Sigma-Aldrich

5β-Pregnane-3β,11β,17α,21-tetrol-20-one

Synonym: 3β,11β,17α,21-Tetrahydroxy-5β-pregnan-20-one, 3β-Tetrahydrocortisol

  • CAS Number 15734-50-6

  • Empirical Formula (Hill Notation) C21H34O5

  • Molecular Weight 366.49

Purchase
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?