EMAIL THIS PAGE TO A FRIEND
P1771 Sigma-Aldrich

L-(+)-Penicillamine

Synonym: 3,3-Dimethyl-L-cysteine, 3-Mercapto-L-valine, L-(+)-2-Amino-3-mercapto-3-methylbutanoic acid

  • CAS Number 1113-41-3

  • Empirical Formula (Hill Notation) C5H11NO2S

  • Molecular Weight 149.21

  •  Beilstein/REAXYS Number 1722374

  •  EC Number 214-203-9

  •  MDL number MFCD00064303

Purchase

Properties

mp   ~200 °C (dec.)
storage temp.   2-8°C
SMILES string   CC(C)(S)[C@H](N)C(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
YV9445500

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

196312 L-Penicillamine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?