• USA Home
  • P1801 - N-Phthaloyl-L-glutamic acid

EMAIL THIS PAGE TO A FRIEND
P1801 Sigma-Aldrich

N-Phthaloyl-L-glutamic acid

Synonym: PhGA

  • CAS Number 340-90-9

  • Empirical Formula (Hill Notation) C13H11NO6

  • Molecular Weight 277.23

  •  Beilstein/REAXYS Number 90136

  •  MDL number MFCD00066476

Purchase

Properties

mp   160-162 °C (lit.)
storage temp.   2-8°C
SMILES string   OC(=O)CC[C@H](N1C(=O)c2ccccc2C1=O)C(O)=O
InChI   1S/C13H11NO6/c15-10(16)6-5-9(13(19)20)14-11(17)7-3-1-2-4-8(7)12(14)18/h1-4,9H,5-6H2,(H,15,16)(H,19,20)/t9-/m0/s1
InChI key   FEFFSKLJNYRHQN-VIFPVBQESA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

330418 N-Phthaloyl-L-glutamic acid, 99% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?