• USA Home
  • P1835 - [Ser25]-Protein Kinase C fragment 19-31

EMAIL THIS PAGE TO A FRIEND
P1835 Sigma

[Ser25]-Protein Kinase C fragment 19-31

≥97% (HPLC)

Purchase

Description

Amino Acid Sequence

Arg-Phe-Ala-Arg-Lys-Gly-Ser-Leu-Arg-Gln-Lys-Asn-Val

Substrates

Protein kinase C substrate, Km = 0.2μM

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?