EMAIL THIS PAGE TO A FRIEND
P1876 Sigma-Aldrich

DL-Phenylalanine

≥98% (TLC)

Synonym: (±)-2-Amino-3-phenylpropionic acid

  • CAS Number 150-30-1

  • Linear Formula C6H5CH2CH(NH2)COOH

  • Molecular Weight 165.19

  •  Beilstein/REAXYS Number 3198807

  •  EC Number 205-756-7

  •  MDL number MFCD00064225

Purchase

Properties

assay   ≥98% (TLC)
mp   266-267 °C (dec.) (lit.)
SMILES string   NC(Cc1ccccc1)C(O)=O
InChI   1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)
InChI key   COLNVLDHVKWLRT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

147966 DL-Phenylalanine, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?