• USA Home
  • P1902 - 1,2-Dipalmitoyl-rac-glycero-3-phospho-L-serine

EMAIL THIS PAGE TO A FRIEND
P1902 Sigma-Aldrich

1,2-Dipalmitoyl-rac-glycero-3-phospho-L-serine

~98%

Synonym: 1,2-Dihexadecanoyl-rac-glycero-3-phospho-L-serine, DL-α-Phosphatidyl-L-serine, dipalmitoyl

  • CAS Number 3036-82-6

  • Empirical Formula (Hill Notation) C38H74NO10P

  • Molecular Weight 735.97

  •  Beilstein/REAXYS Number 2230951

  •  PubChem Substance ID 24898255

  •  NACRES NA.25

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophospholipid Metabolism, Glycerophospholipids, Glycerophosphoserines,
Quality Level   200
assay   ~98%
functional group   phospholipid
shipped in   ambient
storage temp.   −20°C
SMILES string   CCCCCCCCCCCCCCCC(=O)OCC(COP(O)(=O)OC[C@H](N)C(O)=O)OC(=O)CCCCCCCCCCCCCCC
InChI   1S/C38H74NO10P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(40)46-31-34(32-47-50(44,45)48-33-35(39)38(42)43)49-37(41)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h34-35H,3-33,39H2,1-2H3,(H,42,43)(H,44,45)/t34?,35-/m0/s1
InChI key   KLFKZIQAIPDJCW-HTIIIDOHSA-N

Description

Packaging

25 mg in poly bottle

5 mg in glass bottle

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. P1902.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?