EMAIL THIS PAGE TO A FRIEND
P2641 Sigma-Aldrich

2-Phenethylamine

liquid

Synonym: β-Phenylethylamine, 2-Phenylethylamine

  • CAS Number 64-04-0

  • Linear Formula C6H5CH2CH2NH2

  • Molecular Weight 121.18

  •  Beilstein/REAXYS Number 507488

  •  EC Number 200-574-4

  •  MDL number MFCD00008184

  •  PubChem Substance ID 329820144

Purchase

Properties

Related Categories Alkaloid, Cell Biology, Nutrition Research, Phytochemicals by Chemical Classification
vapor density   4.18 (vs air)
InChI Key   BHHGXPLMPWCGHP-UHFFFAOYSA-N
form   liquid
color   clear colorless
refractive index   n20/D 1.533 (lit.)
bp   197-200 °C(lit.)
density   0.962 g/mL at 20 °C (lit.)
storage temp.   2-8°C

Description

Preparation Note

Redistilled

Other Notes

This product has been replaced by 407267-ALDRICH | Phenethylamine purified by redistillation, ≥99.5%

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2922 6.1(8) / PGII
WGK Germany 
1
RTECS 
SG8750000
Flash Point(F) 
177.8 °F
Flash Point(C) 
81 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?