• USA Home
  • P2648 - 5β-Pregnane-3α,11β,17α,20β-tetrol

EMAIL THIS PAGE TO A FRIEND
P2648 Supelco

5β-Pregnane-3α,11β,17α,20β-tetrol

Synonym: 3α,11β,17α,20β-Tetrahydroxy-5β-pregnane

  • CAS Number 2434-06-2

  • Empirical Formula (Hill Notation) C21H36O4

  • Molecular Weight 352.51

Purchase
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?