• USA Home
  • P2673 - N6-(2-Phenylethyl)adenosine

EMAIL THIS PAGE TO A FRIEND
P2673 Sigma-Aldrich

N6-(2-Phenylethyl)adenosine

  • CAS Number 20125-39-7

  • Empirical Formula (Hill Notation) C18H21N5O4

  • Molecular Weight 371.39

  •  MDL number MFCD00055128

  •  PubChem Substance ID 24898349

Purchase

Properties

Related Categories Adenosines/P2 Nucleotide Receptors (Purinergics), Agonists, Cell Biology, Cell Signaling and Neuroscience, Neuroscience,
InChI Key   LGZYEDZSPHLISU-UHFFFAOYSA-N
solubility   0.1 M NaOH: soluble
  H2O: very slightly soluble
  ethanol: moderately soluble
storage temp.   2-8°C

Description

Biochem/physiol Actions

Selective A1 adenosine receptor agonist.

Application

N6-(2-Phenylethyl)adenosine is an adenosine analogue that upon phosphorylation may be used to study the cystic fibrosis transmembrane conductance regulator (G551D-CFTR). N6-(2-Phenylethyl)adenosine is used to study the effects of adenosine analogues on adenosine receptor functions.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?