• USA Home
  • P2756 - 17α,20β-Dihydroxy-4-pregnen-3-one

EMAIL THIS PAGE TO A FRIEND
P2756 Sigma-Aldrich

17α,20β-Dihydroxy-4-pregnen-3-one

Synonym: 17α,20β-Dihydroxyprogesterone, 17α-Hydroxy-20β-dihydroprogesterone, 4-Pregnene-17α,20β-diol-3-one, Maturation Inducing hormone (MIF)

  • CAS Number 1662-06-2

  • Empirical Formula (Hill Notation) C21H32O3

  • Molecular Weight 332.48

  •  MDL number MFCD00010485

Purchase

Properties

SMILES string   O=C1CC[C@@]2(C)C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4(O)[C@@H](C)O)=C1
InChI   1S/C21H32O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h12-13,16-18,22,24H,4-11H2,1-3H3/t13-,16-,17+,18+,19+,20+,21+/m1/s1
InChI key   MASCESDECGBIBB-FSHQYNQFSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?