• USA Home
  • P3121 - Phe-Phe-Phe-Phe-Phe acetate salt

EMAIL THIS PAGE TO A FRIEND
P3121 Sigma-Aldrich

Phe-Phe-Phe-Phe-Phe acetate salt

≥97% (HPLC)

Synonym: Penta-L-phenylalanine

  • CAS Number 65757-10-0

  • Empirical Formula (Hill Notation) C45H47N5O6 · C2H4O2

  • Molecular Weight 813.94

  •  MDL number MFCD00083786

  •  PubChem Substance ID 24898392

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   NC(Cc1ccccc1)C(=O)NC(Cc2ccccc2)C(=O)NC(Cc3ccccc3)C(=O)NC(Cc4ccccc4)C(=O)NC(Cc5ccccc5)C(O)=O
InChI   1S/C45H47N5O6/c46-36(26-31-16-6-1-7-17-31)41(51)47-37(27-32-18-8-2-9-19-32)42(52)48-38(28-33-20-10-3-11-21-33)43(53)49-39(29-34-22-12-4-13-23-34)44(54)50-40(45(55)56)30-35-24-14-5-15-25-35/h1-25,36-40H,26-30,46H2,(H,47,51)(H,48,52)(H,49,53)(H,50,54)(H,55,56)
InChI key   YQUHACPEXSVYDL-UHFFFAOYSA-N

Description

Amino Acid Sequence

Phe-Phe-Phe-Phe-Phe

Packaging

10 mg in glass bottle

50 mg in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?