• USA Home
  • P3142 - 1,2-Dibutyryl-sn-glycero-3-phosphocholine

EMAIL THIS PAGE TO A FRIEND
P3142 Sigma-Aldrich

1,2-Dibutyryl-sn-glycero-3-phosphocholine

20 mg/mL in chloroform: methanol (3:1), solution, ≥99%

Synonym: (7R)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-(1-oxobutoxy)-3,5,9-trioxa-4-phosphatridecan-1-aminium 4-oxide, inner salt, L-α-Phosphatidylcholine, dibutyryl, Dibutanoyllecithin, Dibutyryl lecithin, PC(4:0/4:0)

  • CAS Number 3355-26-8

  • Empirical Formula (Hill Notation) C16H32NO8P

  • Molecular Weight 397.40

  •  MDL number MFCD00050430

  •  PubChem Substance ID 24898395

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophospholipid Metabolism, Glycerophospholipids, Glycerophosphonocholines,
InChI Key   GERHSIUYKQRHRP-UHFFFAOYSA-N
assay   ≥99%
form   solution
concentration   20 mg/mL in chloroform: methanol (3:1)
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?