• USA Home
  • P3643 - Phenobarbital solution

EMAIL THIS PAGE TO A FRIEND
P3643 Supelco

Phenobarbital solution

1.0 mg/mL in methanol

Synonym: 5-Ethyl-5-phenylbarbituric acid solution

  • CAS Number 50-06-6

  • Empirical Formula (Hill Notation) C12H12N2O3

  • Molecular Weight 232.24

  •  MDL number MFCD00066494

  •  PubChem Substance ID 329820184

Purchase

Properties

grade   drug standard
drug control   Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   2-8°C
SMILES string   CCC1(C(=O)NC(=O)NC1=O)c2ccccc2
InChI   1S/C12H12N2O3/c1-2-12(8-6-4-3-5-7-8)9(15)13-11(17)14-10(12)16/h3-7H,2H2,1H3,(H2,13,14,15,16,17)
InChI key   DDBREPKUVSBGFI-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Basic Drugs on Equity®-5

From our library of Articles, Sigma-Aldrich presents GC Analysis of Basic Drugs on Equity®-5
Keywords: Chromatography, Gas chromatography, Purification

GC Analysis of Basic Drugs on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Basic Drugs on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

HPLC Analysis of Barbiturates in Serum on Discovery® C18 after SPE using Discovery® DSC-18Lt

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates in Serum on Discovery® C18 after SPE using Discovery® DSC-18Lt
Keywords: Calorimetry, Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical, Sample preparations, Separation, Solid phase extractions, Solvents

HPLC Analysis of Barbiturates on Ascentis® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Ascentis® C18
Keywords: Anticonvulsants, Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical, Separation, Solvents

HPLC Analysis of Barbiturates on Ascentis® C8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Ascentis® C8
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Barbiturates on Discovery® C18, Unbuffered Mobile Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® C18, Unbuffered Mobile Phase
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Barbiturates on Discovery® C8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® C8
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Barbiturates on Discovery® RP-AmideC16

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® RP-AmideC16
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

TLC Analysis of Drugs on HPTLC Silica Gel 60 RP-18 F254s

From our library of Articles, Sigma-Aldrich presents TLC Analysis of Drugs on HPTLC Silica Gel 60 RP-18 F254s
Keywords: Chromatography, Pharmaceutical, Thin layer chromatography

UHPLC Analysis of Barbiturates on Titan™ C18

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Barbiturates on Titan™ C18
Keywords: Anti-depressants, Anticonvulsants, Chromatography, Clinical, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pharmaceutical, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?