• USA Home
  • P3881 - DL-Phenylalanine hydroxamate

EMAIL THIS PAGE TO A FRIEND
P3881 Sigma-Aldrich

DL-Phenylalanine hydroxamate

  • CAS Number 36207-44-0

  • Empirical Formula (Hill Notation) C9H12N2O2

  • Molecular Weight 180.20

  •  MDL number MFCD00057707

  •  eCl@ss 32160406

  •  PubChem Substance ID 24898464

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   XWQMGESVWMBPAQ-UHFFFAOYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?