• USA Home
  • P3900 - 10-(1-Pyrene)-10-ketodecanoic acid

EMAIL THIS PAGE TO A FRIEND
P3900 Sigma-Aldrich

10-(1-Pyrene)-10-ketodecanoic acid

  • CAS Number 104180-30-5

  • Empirical Formula (Hill Notation) C26H26O3

  • Molecular Weight 386.48

  •  MDL number MFCD00056580

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?