• USA Home
  • P4001 - Phe-Leu amide hydrobromide

EMAIL THIS PAGE TO A FRIEND
P4001 Sigma-Aldrich

Phe-Leu amide hydrobromide

  • CAS Number 108321-16-0

  • Empirical Formula (Hill Notation) C15H23N3O2 · HBr

  • Molecular Weight 358.27

  •  MDL number MFCD00058005

  •  PubChem Substance ID 24898478

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
storage temp.   −20°C
SMILES string   Br.CC(C)CC(NC(=O)C(N)Cc1ccccc1)C(N)=O
InChI   1S/C15H23N3O2.BrH/c1-10(2)8-13(14(17)19)18-15(20)12(16)9-11-6-4-3-5-7-11;/h3-7,10,12-13H,8-9,16H2,1-2H3,(H2,17,19)(H,18,20);1H
InChI key   SBFSMRHQAGKROZ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?