• USA Home
  • P4025 - 10-(1-Pyrene)-10-ketodecanoylcholesterol

EMAIL THIS PAGE TO A FRIEND
P4025 SAFC

10-(1-Pyrene)-10-ketodecanoylcholesterol

~95% (HPLC)

Synonym: 3β-Hydroxy-5-cholestene 3-(10-[1-pyrene]-10-keto)decanoate, 5-Cholesten-3β-ol 3-(10-[1-pyrene]-10-keto)decanoate

  • CAS Number 108321-46-6

  • Empirical Formula (Hill Notation) C53H70O3

  • Molecular Weight 755.12

Purchase

Properties

assay   ~95% (HPLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?