• USA Home
  • P4402 - (S)-(−)-1-Phenylethanol

EMAIL THIS PAGE TO A FRIEND
P4402 Sigma

(S)-(−)-1-Phenylethanol

Synonym: (−)-Methyl phenyl carbinol, (S)-(−)-α-Methylbenzyl alcohol, (S)-(−)-sec-Phenylethyl alcohol

  • CAS Number 1445-91-6

  • Empirical Formula (Hill Notation) C8H10O

  • Molecular Weight 122.16

  •  Beilstein Registry Number 2039797

  •  MDL number MFCD00064264

  •  PubChem Substance ID 24898536

Purchase

Properties

InChI Key   WAPNOHKVXSQRPX-ZETCQYMHSA-N
bp   88-89 °C/10 mmHg(lit.)
mp   9-11 °C (lit.)
density   1.012 g/mL at 20 °C(lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2937 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C
Protocols & Articles

Articles

GC Analysis of 1-Phenylethanol Enantiomers on Astec® CHIRALDEX® B-PM

From our library of Articles, Sigma-Aldrich presents GC Analysis of 1-Phenylethanol Enantiomers on Astec® CHIRALDEX® B-PM
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Related Products

replaced by

Product #

Description

Add to Cart

77849 (S)-(−)-1-Phenylethanol, for chiral derivatization, LiChropur, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?