• USA Home
  • P4613 - Proadrenomedullin Fragment 1-20 human

EMAIL THIS PAGE TO A FRIEND
P4613 Sigma-Aldrich

Proadrenomedullin Fragment 1-20 human

>93% (HPLC), solid

Synonym: PAMP

  • CAS Number 150238-87-2

  • Empirical Formula (Hill Notation) C112H178N36O27

  • Molecular Weight 2460.84

  •  MDL number MFCD01076554

Purchase

Properties

assay   >93% (HPLC)
form   solid
storage temp.   −20°C
Gene Information   human ... ADM(133)

Description

Amino Acid Sequence

Ala-Arg-Leu-Asp-Val-Ala-Ser-Glu-Phe-Arg-Lys-Lys-Trp-Asn-Lys-Trp-Ala-Leu-Ser-Arg-NH2

Biochem/physiol Actions

Precursor of adrenomedullin producing rapid and hypotensive effects and suggested to function as an inhibitory modulator of renal noradrenergic neurotransmission, regulates renal function, inhibitory regulator of adrenal catecholamine release and induces potent hyperglycemic effects.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?