EMAIL THIS PAGE TO A FRIEND
P4764 Sigma-Aldrich

1-Pentadecene

~98%

  • CAS Number 13360-61-7

  • Linear Formula CH3(CH2)12CH=CH2

  • Molecular Weight 210.40

  •  Beilstein/REAXYS Number 1751011

  •  EC Number 236-414-5

  •  MDL number MFCD00008987

Purchase

Properties

InChI Key   PJLHTVIBELQURV-UHFFFAOYSA-N
assay   ~98%
refractive index   n20/D 1.439 (lit.)
bp   268-269 °C (lit.)
mp   −4 °C (lit.)
density   0.775 g/mL at 25 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?