• USA Home
  • P4899 - 1,2-Distearoyl-sn-glycero-3-phosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
P4899 Sigma-Aldrich

1,2-Distearoyl-sn-glycero-3-phosphate sodium salt

≥99%

Synonym: 1,2-Dioctadecanoyl-sn-glycero-3-phosphate sodium salt, L-α-Distearoyl phosphatidic acid sodium salt

  • CAS Number 108321-18-2

  • Empirical Formula (Hill Notation) C39H76NaO8P

  • Molecular Weight 726.98

  •  MDL number MFCD00153888

  •  PubChem Substance ID 24898567

  •  NACRES NA.25

Purchase

Properties

Related Categories Biochemicals and Reagents, Glycerophosphates, Glycerophospholipids, Lipids
Quality Level   200
assay   ≥99%
form   powder
functional group   ester
shipped in   ambient
storage temp.   −20°C
SMILES string   [H][C@@](COP([O-])(O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]
InChI   1S/C39H77O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h37H,3-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/t37-;/m0./s1
InChI key   ALPWRKFXEOAUDR-JFRIYMKVSA-M
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?