EMAIL THIS PAGE TO A FRIEND
P5272 Sigma-Aldrich

N-Phenylthiourea

Grade II, ≥90% (HPLC)

Synonym: 1-Phenyl-2-thiourea, PTU, Phenylthiocarbamide

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thioureas More...
type   Grade II
assay   ≥90% (HPLC)
mp   145-150 °C (lit.)
SMILES string   NC(=S)Nc1ccccc1
InChI   1S/C7H8N2S/c8-7(10)9-6-4-2-1-3-5-6/h1-5H,(H3,8,9,10)
InChI key   FULZLIGZKMKICU-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGI
WGK Germany 
3
RTECS 
YU1400000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?