• USA Home
  • P5626 - 2-(4-Biphenylyl)-5-phenyl-1,3,4-oxadiazole

EMAIL THIS PAGE TO A FRIEND
P5626 Sigma-Aldrich

2-(4-Biphenylyl)-5-phenyl-1,3,4-oxadiazole

suitable for liquid scintillation spectrometry

Synonym: 2-Phenyl-5-(4-biphenylyl)-1,3,4-oxadiazole, PBD

  • CAS Number 852-38-0

  • Empirical Formula (Hill Notation) C20H14N2O

  • Molecular Weight 298.34

  •  Beilstein/REAXYS Number 252554

  •  EC Number 212-712-0

  •  MDL number MFCD00003100

Purchase

Properties

mp   167-169 °C (lit.)
suitability   suitable for liquid scintillation spectrometry
storage temp.   2-8°C
SMILES string   c1ccc(cc1)-c2ccc(cc2)-c3nnc(o3)-c4ccccc4
InChI   1S/C20H14N2O/c1-3-7-15(8-4-1)16-11-13-18(14-12-16)20-22-21-19(23-20)17-9-5-2-6-10-17/h1-14H
InChI key   WMAXWOOEPJQXEB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

257850 2-(4-Biphenylyl)-5-phenyl-1,3,4-oxadiazole, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?