• USA Home
  • P5650 - 1,2-Dihexadecanoyl-rac-glycero-3-phospho-rac-(1-glycerol) ammonium salt

EMAIL THIS PAGE TO A FRIEND
P5650 Sigma-Aldrich

1,2-Dihexadecanoyl-rac-glycero-3-phospho-rac-(1-glycerol) ammonium salt

≥99% (GC)

Synonym: DL-α-Phosphatidyl-DL-glycerol, dipalmitoyl ammonium salt, L-α-Phosphatidyl-DL-glycerol, dipalmitoyl

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophosphoglycerols, Glycerophospholipid Metabolism, Glycerophospholipids,
InChI Key   KSVUSCAQEBSOIJ-UHFFFAOYSA-N
assay   ≥99% (GC)
storage temp.   −20°C

Description

Packaging

100 mg in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?