• USA Home
  • P5652 - Phe-Leu-Arg-Phe amide

EMAIL THIS PAGE TO A FRIEND
P5652 Sigma-Aldrich

Phe-Leu-Arg-Phe amide

≥97% (HPLC)

Synonym: FLRF amide

  • CAS Number 104180-32-7

  • Empirical Formula (Hill Notation) C30H44N8O4

  • Molecular Weight 580.72

  •  MDL number MFCD00076630

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Linkage

Molluscan cardioexcitatory neuropeptide analog

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?