EMAIL THIS PAGE TO A FRIEND
P5771 Sigma-Aldrich

Palmitic anhydride

Grade II, ≥90%, ~99% fatty acid basis

Synonym: Hexadecanoic anhydride

  • CAS Number 623-65-4

  • Linear Formula [CH3(CH2)14CO]2O

  • Molecular Weight 494.83

  •  Beilstein/REAXYS Number 1807507

  •  EC Number 210-805-0

  •  MDL number MFCD00008992

  •  PubChem Substance ID 24898705

Purchase

Properties

Related Categories Anhydrides, Biochemicals and Reagents, Building Blocks, Carbonyl Compounds, Carboxylic Acid Anhydrides,
InChI Key   QWZBEFCPZJWDKC-UHFFFAOYSA-N
type   Grade II
assay   ≥90%
  ~99% fatty acid basis
mp   61-64 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?