• USA Home
  • P6284 - 1-O-Palmityl-2-palmitoyl-rac-glycero-3-phosphocholine

EMAIL THIS PAGE TO A FRIEND
P6284 Sigma-Aldrich

1-O-Palmityl-2-palmitoyl-rac-glycero-3-phosphocholine

~99%

Synonym: DL-α-Phosphatidylcholine, β-hexadecanoyl-γ-O-hexadecyl

  • CAS Number 84743-00-0

  • Empirical Formula (Hill Notation) C40H82NO7P

  • Molecular Weight 720.06

  •  MDL number MFCD00063524

  •  PubChem Substance ID 24898754

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophospholipid Metabolism, Glycerophospholipids, Glycerophosphonocholines,
assay   ~99%
form   solid
functional group   phospholipid
shipped in   ambient
storage temp.   −20°C
SMILES string   CCCCCCCCCCCCCCCCOCC(COP(O)(=O)OCC[N](C)(C)C)OC(=O)CCCCCCCCCCCCCCC
InChI   1S/C40H83NO7P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-35-45-37-39(38-47-49(43,44)46-36-34-41(3,4)5)48-40(42)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h39H,6-38H2,1-5H3,(H,43,44)
InChI key   HWWCVCRAHSNKKC-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?