• USA Home
  • P6292 - DL-Pantothenic acid hemicalcium salt

EMAIL THIS PAGE TO A FRIEND
P6292 Sigma-Aldrich

DL-Pantothenic acid hemicalcium salt

suitable for cell culture, suitable for insect cell culture

Synonym: (±)-N-(2,4-Dihydroxy-3,3-dimethylbutyryl)-β-alanine, DL-N-(2,4-Dihydroxy-3,3-dimethylbutyryl)-β-alanine, Calcium pantothenate

  • CAS Number 63409-48-3

  • Linear Formula HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 · 1/2Ca2+

  • Molecular Weight 238.27

  •  MDL number MFCD00063189

  •  PubChem Substance ID 24898756

Purchase

Properties

Related Categories Amino Acids and Vitamins, Cell Culture, Companion Products and Reagents, Insect Platform, Reagents and Supplements,
InChI Key   FAPWYRCQGJNNSJ-UHFFFAOYSA-L
product line   BioReagent
form   powder
mol wt   Mw 238.27 g/mol
application(s)   cell culture | insect: suitable
  cell culture | mammalian: suitable
  cell culture | plant: suitable
color   white
mp   138 °C (dec.) (lit.)
solubility   water: soluble 50 mg/mL, clear to slightly hazy, colorless to faintly yellow
storage temp.   2-8°C

Description

Application

Cell culture media are generally supplemented with D-Pantothenic acid (vitamin B5), an essential vitamin required for the synthesis of coenzyme-A (CoA) and the metabolism of proteins, carbohydrates, and fats.

This product has been qualified for use in plant cell culture. Also see P5155 for D-Pantothenic acid qualified for use in animal, insect and plant cell culture.

Biochem/physiol Actions

A vitamin that supports the synthesis of carbohydrates, proteins and fats through formation of coenzyme-A (CoA).

Caution

This product is a mixture of D and L isomers. Only the dextrorotatory (D) isomer of pantothenic acid possesses biologic activity. The levorotatory (L) form may antagonize the effects of the dextrorotatory isomer.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Pipette Controller from Brand
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Related Products

comparable product

Product #

Description

Add to Cart

P5155 D-Pantothenic acid hemicalcium salt, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?